Learn More
Proteintech FlexAble FITC Plus Antibody Labeling Kit for Human IgG
FlexAble FITC Plus Antibody Labeling Kit for Human IgG is a novel antibody labeling kit that uses an affinity linker to conjugate FITC Plus in any buffer condition to Human IgG primary antibodies from any supplier.
Marque:  Proteintech KFA109-200
Informations supplémentaires : Poids : 0.02000kg
 Afficher plus de versions de ce produit
Code produit. 18030596
  • 1059.97€ / 200 réactions
En stock
pour voir le stock disponible 
Description
Description
FlexAble FITC Plus Antibody Labeling Kit for Human IgG is a novel antibody labeling kit that uses an affinity linker to conjugate FITC Plus in any buffer condition to Human IgG primary antibodies from any supplier. One labeling reaction requires only 0.5 μg of antibody - regardless of the antibody concentration. Label your Human IgG antibody in<10 min without any additional equipment.
Spécification
Spécification
100 Tests