Learn More
Proteintech FlexAble CoraLite Plus 488 Antibody Labeling Kit for Human IgG
FlexAble CoraLite Plus 488 Antibody Labeling Kit for Human IgG is a novel antibody labeling kit that uses an affinity linker to conjugate CoraLite Plus 488 in any buffer condition to Human IgG primary antibodies from any supplier.
Marque:  Proteintech KFA104-10
Informations supplémentaires : Poids : 0.02000kg
Code produit. 18024235
  • 107.31€ / 10 réactions
En stock
pour voir le stock disponible 
Description
Description
FlexAble CoraLite™ Plus 488 Antibody Labeling Kit for Human IgG is a novel antibody labeling kit that uses an affinity linker to conjugate CoraLite™ Plus 488 in any buffer condition to Human IgG primary antibodies from any supplier. One labeling reaction requires only 0.5 μg of antibody - regardless of the antibody concentration. Label your Human IgG antibody in<10 min without any additional equipment.
Spécification
Spécification
10 Tests