missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FITC Mouse, Clone: 3C10A1, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 68132-1-IG-150UL
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.00010kg
Description
FITC Monoclonal antibody specifically detects FITC in fitc samples. It is validated for ELISASpécification
FITC | |
Monoclonal | |
1000 μg/mL | |
PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
Mouse | |
150 μL | |
Store at -20°C | |
Liquid |
ELISA | |
3C10A1 | |
Unconjugated | |
BC002899 | |
Protein A purification | |
Primary | |
Antibody | |
IgG2a |