Learn More
Evocalcet, MedChemExpress
MedChemExpress Evocalcet has an activating effect on calcium sensing receptor (CaSR) extracted from patent WO 2017061621 A1, compound A.
Marque: Medchem Express HY-17613
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Evocalcet has an activating effect on calcium sensing receptor (CaSR) extracted from patent WO 2017061621 A1, compound A.Spécification
Evocalcet | |
Off-White | |
C24H26N2O2 | |
KHK7580 | |
O=C(O)CC1=CC=C(N2C[C@@H](N[C@@H](C3=C4C=CC=CC4=CC=C3)C)CC2)C=C1 | |
374.48 | |
Research | |
Metabolism-protein/nucleotide metabolism |
870964-67-3 | |
Research | |
50 mg | |
DMSO : 30 mg/mL (80.11 mM; Need ultrasonic and warming) | |
374.48 | |
99.05% | |
Solid |
Research purposes only