Learn More
Karl Hecht™ Flacons à fond rond en verre borosilicaté DURAN™
For the safe and easy mixing, pouring and storage of chemicals in the laboratory. Karl Hecht™ Assistent™ DURAN™ Round Bottom Flasks are manufactured from durable DURAN borosilicate 3.3 glass.
Marque:  Karl Hecht™ 41913010
Code produit. 10217191
  • 17.00€ / 1 pièce
Découvrez plus d'offres spéciales
Description
Description
  • Without stopper
  • With standard ground
  • Reinforced ground neck marked with standard ground size
Spécification
Spécification
50 mL
Borosilicate Glass, Duran Glass
Rond
NS 19/26