Learn More
Abnova™ Deoxyribonuclease I (Bovine), Molecular Biology Grade
Used for Enzyme
Marque: Abnova™ P5309.100Units
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
#REF!Spécification
P00639 | |
Enzyme Activity | |
282217 | |
Deoxyribonuclease I (Bovine), Molecular Biology Grade | |
100 U | |
RUO | |
DNASE1 | |
≥2,000 Kunitz units per ml. One Unit causes an increase in absorbance at 260nm of 0.001 per minute per ml at 25°C when acting upon highly polymerized DNA at pH 5.0. | |
None | |
Liquid |
∼ 1 mg/mL | |
In 50% glycerol and 1mM calcium chloride | |
31.3kDa | |
Native protein chromatographically purified from Bovine Pancreas | |
Store at -20°C. | |
DNASE1 | |
Deoxyribonuclease I Assay; Nick translation; DNA mapping; Isolation of nuclear RNA and protein; Plasmid construction; RNA polymerase synthesis of RNA probes and RT-PCR. | |
Bovine | |
Native protein chromatographically purified from Bovine Pancreas |
Correction du contenu d'un produit
Nom du produit
Abnova™ Deoxyribonuclease I (Bovine), Molecular Biology Grade > 100 Units
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu