Learn More
Cytiva Portoirs complets pour tubes à essai
À utiliser avec le collecteur de fractions F9-R et le collecteur de fractions 920
Brand:  Cytiva 18-3050-03
 View more versions of this product
Product Code. 11320272
  • 737.00€ / 1 pièce
En stock
pour voir le stock disponible 
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
Comprend un bol, un support de tube, un socle et un guide. À utiliser avec le collecteur de fractions F9-R et le collecteur de fractions 920.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Comprend un bol, un support de tube, un socle et un guide
Tubes (18 mm)
À utiliser avec le collecteur de fractions F9-R et le collecteur de fractions 920
18 mm
Portoir pour tubes à essai
95
1/boîte
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.