Learn More
CD45RB Antibody (BRA-11 (same as BRA-11G)), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NBP2-34564AF350
429.60 EUR valable jusqu'au 2025-03-28
Utilisez le code promo "25306" pour bénéficier de cette offre.
Descrizione
CD45RB Monoclonal antibody specifically detects CD45RB in Human, Monkey (Negative), Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
Specifica
CD45RB | |
Monoclonal | |
Alexa Fluor 350 | |
B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
Non-T, non-B CALLA positive ALL cell line REH (Leucocyte Workshop IV and V) | |
0.1 mL | |
Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Microglia Markers, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
5788 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
BRA-11 (same as BRA-11G) | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Monkey (Negative), Rat (Negative) | |
Purified |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.