Learn More
Wesbart™ Cardboard Cryoboxes
Wesbart cryogenic boxes are made from fibreboard cardboard and are water-repellent for longer durability.
Marque: Wesbart™ A1/100
Informations supplémentaires : Poids : 1.00000kg
Description
- Cardboard boxes are specially made for the aluminum freezer racks.
- Dividers are available in different diameters.
Spécification
Cardboard Box with Dividers | |
Cardboard | |
Cryo Box | |
Grid Divider | |
10 x 10 | |
50 mm |
White | |
Rectangle | |
100 Cryovials (1 to 2 mL) | |
Non-sterile | |
133 x 133 x 50 mm |