Learn More
Thermo Scientific™ CaptureSelect™ FSH Ligand Leakage ELISA
Thermo Scientific™ CaptureSelect™ FSH Ligand Leakage ELISA is designed to detect possible leached ligand of follicle-stimulating hormone (FSH) purified using CaptureSelect FSH Affinity Matrix.
Marque: Thermo Scientific™ 810318010
Description
CaptureSelect FSH Ligand Leakage ELISA is designed to detect possible leached ligand of follicle-stimulating hormone (FSH) purified using CaptureSelect FSH Affinity Matrix. The ligand, a 14 kD recombinant protein, is coupled by multipoint attachment through stable amide bonds to give high chemical stability and low leakage. Even when covalently attached, small amounts of the affinity ligand can leach off of the chromatography support and co-elute with the protein.
This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect FSH Ligand Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.
Spécification
Store in freezer (-5°C to -30°C). | |
Microplate Reader | |
10 Assays | |
Kit |
ELISA | |
CaptureSelect™ | |
ELISA Kit |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.