Learn More
Robu™ Büchner Filter Funnels with Porosity 2 VitraPOR™ Glass Filter Discs
Robu™ Büchner Filter Funnels with Glass Filter Discs are made of pure borosilicate 3.3 glass and are highly resistant to most chemicals, including solvents, acids, and alkalis. The VitraPOR™ filter disc is rated as Porosity 2 (Pore size: 40 to 100 μm, Standard: ISO 4793-80).
Marque: Robu™ 21122
Informations supplémentaires : Poids : 0.10000kg
Description
- Product Type: Büchner Filter Funnel
- Material: Borosilicate 3.3 Glass (Body), VitraPOR™ Glass (Filter)
- Porosity: Grade 2
- Pore Size: 40-100 μm
- Standard: ISO 4793-80
- For Use With (Application): Filtration
- For Use With (Equipment): Guko Rubber Gaskets (e.g. 10430652)
Spécification
65mm | |
10 mm | |
56 mm | |
Très résistant à la plupart des produits chimiques tels que les solvants, les acides et les alcalins | |
Guko Rubber Gaskets (e.g. 10430652) | |
4.2 oz. |
125 ml | |
Verre borosilicaté | |
ISO 4793-80 | |
65 mm | |
40 to 100 μm | |
Entonnoir de filtration Buchner |