Learn More
STEMCELL Technologies BloodStor™ 100
Biopreservation reagent for cells and tissues
Marque: STEMCELL Technologies 07938
Informations supplémentaires : Poids : 0.50000kg
Description
- BloodStor™ is a series of generic cGMP freezing media products used to cryopreserve stem cells and other cells isolated from umbilical cord blood, peripheral blood, bone marrow, and other biologics
- BloodStor™ 100 contains 100% (w/v) DMSO USP
Spécification
Hematopoietic Stem and Progenitor Cells | |
Cryopreservation | |
5 x 100 mL |