Learn More
BRAND™ Pipette de mesure en verre de classe AS AR Blaubrand™ Eterna
All BLAUBRAND™ graduated pipettes are supplied with a batch number and an accompanying batch certificate in the original packaging. On request, they are also available with an individual certificate, USP individual certificate or DAkkS calibration certificate.
Marque: BRAND™ 27845
Code nomenclature Nacres: NB.05
Informations supplémentaires : Poids : 0.21000kg
Description
Pipette graduée en verre, Type 2, classe AS• DIN EN ISO 835
• Certificat de lot
• Volume nominal en haut
• Ecoulement total
• Verre sodocalcique
Spécification
Cotton Plug Upper End | |
25 mL |