Learn More
IL-8 Mouse anti-Human, Unlabeled, Clone: G265-8, BD
Mouse Monoclonal Antibody
Marque: BD Biosciences 554717
132.68 EUR valable jusqu'au 2025-12-31
Utilisez le code promo "25339" pour bénéficier de cette offre.
Description
The G265-8 antibody reacts with both the 72 and 77 amino acid forms of human interleukin-8 (IL-8). The immunogen used to produce the G265-8 hybridoma was E. coli-expressed recombinant human IL-8.

Spécification
IL-8 | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Recombinant Human IL-8 | |
0.1 mg | |
Immunology | |
Human | |
IgG2b |
Blocking Assay, Flow Cytometry | |
G265-8 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
For Research Use Only.