Learn More
Hamilton™ 50 μL 700 Series Microliter Syringe
The 700 Series Microliter Syringe fulfi the general liquid handling requirements of manufacturing and research laboratories.
Marque:  Hamilton™ CAL80530
Code produit. 17824342
  • 349.00€ / 1 pièce
Découvrez plus d'offres spéciales
Description
Description
  • The 700 series is the original Hamilton syringe.
  • This series of Microliter syringes is designed to solve the general liquid handling requirements of manufacturing and research laboratories.
  • The 700 series remains the industry standard.
  • The plungers and barrels are not interchangeable or replaceable.
  • Dispenses volumes up to 50 μL.
Spécification
Spécification
Non autoclavable
22s G
0,0059 in
0,0283 in
1.03 mm
0.72 mm
122°F
50°F
10°C
50 μl
Aigu, biseauté, courbe, non perforant
Stainless Steel
0,041 in
0.15 mm
0.305 in.
7.75 mm
2 in
50°C
705 N
1
51 mm
Cemented