Learn More
Hamilton™ 50 μL 1700 Series Gas Tight Syringe
The 1700 Series syringes are the Gas Tight version of the 700 series syringes.
Marque:  Hamilton™ CAL84856
Code produit. 17814352
  • 356.00€ / 1 pièce
Découvrez plus d'offres spéciales
Description
Description
  • This series of syringes are ideal for dispensing volumes up to 50 μL.
  • A variety of different terminations and needle options are available in this series.
Spécification
Spécification
Non autoclavable
22s G
0,0059 in
0,0283 in
1.03 mm
0.72 mm
Gas Tight Syringe
50°C
1705 N Syringe
1
51 mm
Cemented
Stainless Steel
0,041 in
0.15 mm
0.305 in.
7.75 mm
2 in
122°F
50°F
10°C
50 μl
Blunt