Learn More
5-cyano-4-phényl-2-[4-(trifluorométhyl)phényl]thiazole, 97 %, Thermo Scientific™
331.00€ - 1321.00€
Identifiants chimiques
CAS | 1858240-54-6 |
---|---|
Formule moléculaire | C17H9F3N2S |
Poids moléculaire (g/mol) | 330.33 |
Numéro MDL | MFCD22682993 |
Clé InChI | XUFKMNGEUDCIIV-UHFFFAOYSA-N |
Synonyme | 4-phenyl-2-4-trifluoromethyl phenyl-1,3-thiazole-5-carbonitrile, 5-cyano-4-phenyl-2-4-trifluoromethyl phenyl thiazole |
CID PubChem | 98000107 |
Nom IUPAC | 4-phényl-2 -[4 -(trifluorométhyl)phényl]-1,3 -thiazole-5 -carbonitrile |
SMILES | FC(F)(F)C1=CC=C(C=C1)C1=NC(=C(S1)C#N)C1=CC=CC=C1 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Identifiants chimiques
1858240-54-6 | |
330.33 | |
XUFKMNGEUDCIIV-UHFFFAOYSA-N | |
98000107 | |
FC(F)(F)C1=CC=C(C=C1)C1=NC(=C(S1)C#N)C1=CC=CC=C1 |
C17H9F3N2S | |
MFCD22682993 | |
4-phenyl-2-4-trifluoromethyl phenyl-1,3-thiazole-5-carbonitrile, 5-cyano-4-phenyl-2-4-trifluoromethyl phenyl thiazole | |
4-phényl-2 -[4 -(trifluorométhyl)phényl]-1,3 -thiazole-5 -carbonitrile |
Spécification
5-Cyano-4-phenyl-2-[4-(trifluoromethyl)phenyl]thiazole | |
C17H9F3N2S | |
UN3439 | |
XUFKMNGEUDCIIV-UHFFFAOYSA-N | |
4-phényl-2 -[4 -(trifluorométhyl)phényl]-1,3 -thiazole-5 -carbonitrile | |
98000107 | |
97% |
1858240-54-6 | |
MFCD22682993 | |
4-phenyl-2-4-trifluoromethyl phenyl-1,3-thiazole-5-carbonitrile, 5-cyano-4-phenyl-2-4-trifluoromethyl phenyl thiazole | |
FC(F)(F)C1=CC=C(C=C1)C1=NC(=C(S1)C#N)C1=CC=CC=C1 | |
330.33 | |
330.33 |
Sécurité et traitement
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Température ambiante
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.