Learn More
Hamilton™ 25 μL 1700 Series Gas Tight Syringe
The 1700 Series syringes are the Gas Tight version of the 700 series syringes.
Marque: Hamilton™ CAL80265
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
- This series of syringes are ideal for dispensing volumes up to 25 μL.
- A variety of different terminations and needle options are available in this series.
Spécification
Non autoclavable | |
22s G | |
0,0059 in | |
0,0283 in | |
0.729 mm | |
0.72 mm | |
Gas Tight Syringe | |
115°C | |
1702 Syringe | |
1 | |
51 mm | |
Amovible |
Borosilicate Glass | |
0.029 in. | |
0.15 mm | |
0.305 in. | |
7.75 mm | |
2 in | |
239°F | |
50°F | |
10°C | |
25 μl | |
Aigu, biseauté, courbe, non perforant |