Learn More
Hamilton™ 10 μL 1700 Series Gas Tight Syringe
The 1700 Series syringes are the Gas Tight version of the 700 series syringes.
Marque: Hamilton™ CAL80011
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
- This series of syringes are ideal for dispensing volumes up to 10 μL.
- A variety of different terminations and needle options are available in this series.
Spécification
Non autoclavable | |
26s G | |
0,005 in | |
0.0187 in. | |
0.460 mm | |
0.47 mm | |
Gas Tight Syringe | |
50°C | |
1701 N | |
1 | |
51 mm | |
Cemented |
Stainless Steel | |
0.018 in. | |
0.13 mm | |
0.260 in. | |
6.60 mm | |
2 in | |
122°F | |
50°F | |
10°C | |
10 μl | |
Aigu, biseauté, courbe, non perforant |