missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNHIT6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA539557
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.00900kg
Spécification
ZNHIT6 | |
Polyclonal | |
Unconjugated | |
ZNHIT6 | |
BCD1, C1orf181 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
229937, 54680 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9NWK9, Q3UFB2 | |
ZNHIT6 | |
A synthetic peptide derived from the internal region of human CA181 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |