Learn More
Cytiva Système Minifold I Spot-Blot complet à 96 puits (acrylique)
Click to view available options
Type:
Minifold I Spot-Blot System
Conditionnement:
1 pièce
Description
- Easy-to-assemble system.
- Available in three interchangeable configurations for spot-, dot-, and slot-blots.
- Accepts all types of transfer membranes: Allows for choice of membrane with the highest binding capacity.
Spécification
Spécification
| Type | Minifold I Spot-Blot System |
| Comprend | Acrylic 96-well manifold apparatus, 5 x Protran BA85 sheets, 5 x 3mm Chr sheets |
| Matériau | Acrylique |
| Volume (métrique) d’échantillon | 200 μL/well |