Learn More
WASL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 14306-1-AP-20UL
Code nomenclature Nacres: NA.46
 Afficher plus de versions de ce produit
Code produit. 16872884
  • 149.35€ / 20µL
Description
The Wiskott-Aldrich syndrome (WAS) family of proteins share similar domain structure, and are involved in transduction of signals from receptors on the cell surface to the actin cytoskeleton. The presence of a number of different motifs suggests that they are regulated by a number of different stimuli, and interact with multiple proteins. Recent studies have demonstrated that these proteins, directly or indirectly, associate with the small GTPase, Cdc42, known to regulate formation of actin filaments, and the cytoskeletal organizing complex, Arp2/3. The WASL gene product is a homolog of WAS protein, however, unlike the latter, it is ubiquitously expressed and shows highest expression in neural tissues. It has been shown to bind Cdc42 directly, and induce formation of long actin microspikes.
Spécification
Spécification
WASL
Polyclonal
Unconjugated
WASL
N WASP, NWASP, WASL, Wiskott Aldrich syndrome like
Rabbit
Antigen Affinity Chromatography
RUO
682507, 73178, 8976
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin)
0.26 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O00401, O08816, Q91YD9
Wasl
WASL Fusion Protein Ag5424
20 μL
Primary
Rat, Mouse, Human