Learn More
ATPG Antibody (OTI2H6), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NBP2-70233AF350
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
ATPG Monoclonal specifically detects ATPG in Human, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Spécification
| ATPG | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| ATP5F1C | |
| Full length recombinant protein of human ATPG (NP_001001973) produced in E.coli. | |
| 0.1 mL | |
| 509.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI2H6 | |
| Western Blot, Immunohistochemistry | |
| ATP synthase gamma chain, mitochondrial, ATP synthase, H+ transporting, mitochondrial F1 complex, gamma polypeptide 1, ATP5CATP synthase subunit gamma, mitochondrial, ATP5CL1mitochondrial ATP synthase, gamma subunit 1, F-ATPase gamma subunit | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Monkey | |
| IgG1 |