Learn More
Abnova™ Human FOXO1A Partial ORF (NP_002006, 556 a.a. - 655 a.a.) Recombinant Protein with GST-tag at N-terminal
Click to view available options
Quantité:
10 ug
25 ug
Conditionnement:
10µg
25µg
Description
This gene belongs to the forkhead family of transcription factors which are characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in myogenic growth and differentiation. Translocation of this gene with PAX3 has been associated with alveolar rhabdomyosarcoma. [provided by RefSeq]
Sequence: TQVKTPVQVPLPHPMQMSALGGYSSVSSCNGYGRMGLLHQEKLPSDLDGMFIERLDCDMESIIRNDLMDGDTLDFNFDNVLPNQSFPHSVKTTTHSWVSG
Spécification
Spécification
| Numéro d’adhésion | NP_002006 |
| À utiliser avec (application) | Antibody Production, ELISA, Protein Array, Western Blot |
| Formule | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
| Identification génétique (Entrez) | 2308 |
| Poids moléculaire | 36.74kDa |
| Nom | FOXO1A (Human) Recombinant Protein (Q01) |
| Test du contrôle qualité | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Quantité | 10 ug |
| Immunogène | TQVKTPVQVPLPHPMQMSALGGYSSVSSCNGYGRMGLLHQEKLPSDLDGMFIERLDCDMESIIRNDLMDGDTLDFNFDNVLPNQSFPHSVKTTTHSWVSG |
| Conditions de stockage | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Afficher plus |