Learn More
Lovibond™ ValidCheck Chlorine
Creating a Primary Chlorine Standard has never been easier! This self-contained ValidCheck Chlorine kit will allow for preparation of 100 mL of 1.5 mg/L NIST traceable primary chlorine standard without need for complicated dilutions or the additional use of pipettes or glassware.
Marque:  Lovibond™ 48105510
Code produit. 17884892
  • 31.90€ / 1 pièce
Expédition estimée: 04-02-2025
pour voir le stock disponible 
Découvrez plus d'offres spéciales
Ajouter au panier
Description
Description
  • Ready-to-use solutions
  • Exactly adjusted concentrations
  • Adapted to special applications
  • Multistandards with top-up solution
Spécification
Spécification
1.5 mg/L Chlorine Standard