missing translation for 'onlineSavingsMsg'
Learn More

UBA6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00€ - 410.00€

Spécification

Antigène UBA6
Concentration 0.27 mg/mL
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugué Unconjugated
Consulter d'autres spécifications

Produits 2
Code produit Marque Quantité Prix Quantité et disponibilité  
Code produit Marque Quantité Prix Quantité et disponibilité  
16801324
Afficher les documents
Proteintech
13211-1-AP-20UL
20 μL
145.00€
20µL
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
16891314
Afficher les documents
Proteintech
13211-1-AP-150UL
150 μL
410.00€
150µL
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
Description

Description

This antibody recognizes the full-length (118 kDa) and the truncated isoform CRA_b (60-70 kDa) of UBA6 (NCBI).

DNA replication, recombination and repair, all of which are necessary for genomic stability, require the presence of exonucleases. In DNA replication, these enzymes are involved in the processing of Okazaki fragments, whereas in DNA repair they function to excise damaged DNA fragments and correct recombinational mismatches. These exonucleases include the family of DNA polymerases. DNA pol alpha, beta, delta and epsilon are involved in DNA replication and repair. DNA pol delta and DNA pol epsilon are multisubunit enzymes, with DNA pol delta consisting of two subunits: p125, which interacts with the sliding DNA clamp protein PCNA; and p50. The nuclear-encoded DNA pol gamma is the only DNA polymerase required for the replication of the mitochondrial DNA. DNA pol zeta is ubiquitously expressed in various tissues and mediates the cellular mechanism of damage-induced mutagenesis. DNA pol theta is a DNA polymerase-helicase that binds ATP and is involved in the repair of interstrand crosslinks.
Spécification

Spécification

UBA6
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
Rabbit
Human, Mouse, Rat
A0AVT1, Q8C7R4
231380, 305268, 55236
UBA6 Fusion Protein Ag4000
Primary
-20°C
UBA6
0.27 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
E1 L2, Monocyte protein 4, MOP 4, MOP4, UBA6, UBE1L2, Ubiquitin activating enzyme 6
UBA6
IgG
Antigen Affinity Chromatography
Antibody
Vidéos
FDS
Documents

Documents

Product Certifications