Learn More
Eppendorf™ twin.tec™ PCR Plates BioBased
Tous les produits Eppendorf Produits
Click to view available options
Couleur:
Incolore
Transparent
Vert
Type de jupe:
À demi-jupe
À jupe
Type de produit:
Plaques PCR BioBased
Plaques PCR LoBind BioBased
Nbre de puits:
96
384
Conditionnement:
25 pièces
Description
- Reduction of consumable-related carbon footprint in the lab
- Laser-engraved lot number and expiration date on each single plate to improve traceability in your laboratory processes
- Unique laser-engraved optical guiding grid and OptiTrack matrix for quick orientation when pipetting manually
- One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance
- Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample
- Exceptionally solid, torque- and heat-resistant polycarbonate frame: No warping, bending or breaking during heat incubation or centrifugation
- Raised well rims for effective sealing, also reduces risk of cross-contamination
- Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean)
- Available with barcode (upon request)
Spécification
Spécification
| Code-barres | Custom Barcoding Available |
| Gamme de produits | Eppendorf |
| Couleur | Incolore |
| Type de produit | Plaques PCR BioBased |
| Type de jupe | À jupe |
| Matériau | Polypropylene, Polycarbonate |
| Nbre de puits | 384 |
| Stérilité | Non stérile |
| Type de puits | Conical Bottom (V-well) |
| Quantité | 25 Pk. |
| Afficher plus |