Learn More
Eppendorf™ twin.tec™ PCR Plates BioBased
Découvrez les nouvelles plaques PCR BioBased twin.tec en polymère biosourcé (approche mass balance). Ces plaques vous permettent de réduire significativement votre empreinte carbone liée aux consommables tout en conservant des performances techniques identiques.
Marque: Eppendorf™ 0030531078
280.00 EUR valable jusqu'au 2025-12-31
Utilisez le code promo "25339" pour bénéficier de cette offre.
Description
- Reduction of consumable-related carbon footprint in the lab
- Laser-engraved lot number and expiration date on each single plate to improve traceability in your laboratory processes
- Unique laser-engraved optical guiding grid and OptiTrack matrix for quick orientation when pipetting manually
- One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance
- Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample
- Exceptionally solid, torque- and heat-resistant polycarbonate frame: No warping, bending or breaking during heat incubation or centrifugation
- Raised well rims for effective sealing, also reduces risk of cross-contamination
- Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean)
- Available with barcode (upon request)
Spécification
Custom Barcoding Available | |
Eppendorf | |
BioBased LoBind PCR Plates | |
Polypropylene, Polycarbonate | |
Non stérile | |
25 Pk. |
LoBind | |
Colorless | |
À jupe | |
384 | |
Conical Bottom (V-well) | |
PCR Ready |