missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tryptase (Mast Cell Marker) Mouse, Clone: TPSAB1/1961, enQuireBio™
Mouse Monoclonal Antibody
199.00€ - 485.00€
Spécification
Antigène | Tryptase (Mast Cell Marker) |
---|---|
Clone | TPSAB1/1961 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15931018
|
EnquireBio
7177-MSM1-P0 |
20 μg | |||||||
15941018
|
EnquireBio
7177-MSM1-P1 |
100 μg |
485.00€
100µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Spécification
Tryptase (Mast Cell Marker) | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant human Tryptase protein fragment (apprx. aa 115-233) | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
31-36kDa |
TPSAB1/1961 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
7177 | |
IgG1 κ | |
Protein A/G purified from Bioreactor concentrate. | |
Secreted and Cytoplasmic | |
Skin or Tonsil |