missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM29 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 17542-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The protein encoded by this gene belongs to the TRIM protein family. It has multiple zinc finger motifs and a leucine zipper motif. It has been proposed to form homo- or heterodimers which are involved in nucleic acid binding. Thus, it may act as a transcriptional regulatory factor involved in carcinogenesis and/or differentiation. It may also function in the suppression of radiosensitivity since it is associated with ataxia telangiectasia phenotype.Spécification
TRIM29 | |
Polyclonal | |
Unconjugated | |
TRIM29 | |
ATDC, TRIM29, tripartite motif containing 29 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23650, 300656, 72169 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q14134, Q8R2Q0 | |
Trim29 | |
TRIM29 Fusion Protein Ag11600 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |