missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tnfaip6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA562259
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Immunogen sequence: DDPGCLADYV EIYDSYDDVH GFVGRYCGDE LPDDIISTGN VMTLKFLSDA SVTAGGFQIK YVAMDPVSKS SQ Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 90%.
Tnfaip6 Polyclonal antibody specifically detects Tnfaip6 in Human samples. It is validated for Immunohistochemistry, Western Blot.
Spécification
| Tnfaip6 | |
| Polyclonal | |
| Unconjugated | |
| TNFAIP6 | |
| Hyaluronate-binding protein; TNF alpha induced protein 6; TNF alpha-induced protein 6; Tnfaip6; Tnfip6; TNF-stimulated gene 6 protein; TSG6; TSG-6; tumor necrosis factor alpha induced protein 6; tumor necrosis factor alpha-induced protein 6; tumor necrosis factor alpha-inducible protein 6; tumor necrosis factor induced protein 6; tumor necrosis factor, alpha induced protein 6; tumor necrosis factor, alpha-induced protein 6; tumor necrosis factor-inducible gene 6 protein; tumor necrosis factor-stimulated gene-6 protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7130 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P98066 | |
| TNFAIP6 | |
| Recombinant protein corresponding to Human Tnfaip6 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu