missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM176B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 19825-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
TMEM176B (also known as LR8) is a transmembrane protein belongs to the MS4A family of proteins. It may play a role in the process of maturation of dendritic cells. TMEM176B was first discovered in human lung fibroblasts and is associated with human small cell lung carcinoma. Studies suggest that TMEM176B might be involved in cancer and might serve as targets for antiangiogenic therapy of cancer patients. There are two isoforms produced by alternative splicing.Spécification
TMEM176B | |
Polyclonal | |
Unconjugated | |
TMEM176B | |
LR8, Protein LR8, TMEM176B, transmembrane protein 176B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
28959, 65963 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3YBM2, Q9R1Q6 | |
TMEM176B | |
TMEM176B Fusion Protein Ag13918 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |