missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16564-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Plays a role in mitochondrial morphogenesis.Spécification
TMEM11 | |
Polyclonal | |
Unconjugated | |
TMEM11 | |
C17orf35, PM1, PMI, Protein PM1, TMEM11, transmembrane protein 11 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
216821, 303196, 8834 | |
-20°C | |
Liquid |
Western Blot | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
B0BN86, P17152, Q8BK08 | |
TMEM11 | |
TMEM11 Fusion Protein Ag9839 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |