missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIM-3 (CD366) Mouse anti-Human, Alexa Fluor 488, Clone: 7D3, BD Biosciences™
Mouse Monoclonal Antibody
202.00€ - 490.00€
Spécification
Antigène | TIM-3 (CD366) |
---|---|
Clone | 7D3 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugué | Alexa Fluor 488 |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
17257814
|
BD Biosciences
567031 |
25 Tests |
202.00€
100µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
17247814
|
BD Biosciences
567030 |
100 Tests |
490.00€
100µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
TIM-3 (CD366) Monoclonal antibody specifically detects TIM-3 (CD366) in Human samples. It is validated for Flow cytometrySpécification
TIM-3 (CD366) | |
Flow Cytometry | |
Alexa Fluor 488 | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human TIM-3 | |
Primary |
7D3 | |
Monoclonal | |
Mouse | |
Human | |
CD366; HAVCR2; TIM3; T cell immunoglobulin mucin-3; TIMD-3; KIM-3 | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor 488 under optimum conditions, and unreacted Alexa Fluor 488 was removed. |