Learn More
Thermo Scientific™ Thermo Scientific TSU Series Undercounter Refrigerators
Tous les produits Thermo Scientific Produits
Click to view available options
Modèle de porte:
Plein
Verre
Type de bouchon:
UE
Conditionnement:
1 pièce
Description
Thermo Scientific TSU Series Undercounter Laboratory Refrigerators offer a compelling cold storage option for a wide variety of laboratories. With a full suite of alarms, whisper quiet operation, and NSF 456 certified performance, the TSU Series is designed specifically for laboratory settings.
- Remote alarm contacts
- NSF 456 Certified
- Whisper quiet operation
- ENERGYStar certified
- Green refrigerants support global greenhouse gas emission reduction initiatives
- Hi/Lo temperature, door ajar, and power fail alarms
Spécification
Spécification
| Capacité (impériale) | 4.5 cu.ft. |
| Capacité (métrique) | 127 L |
| Gamme de températures | 2°C to 8°C |
| Modèle de porte | Plein |
| No. of Doors | 1 |
| Étagères | 2 Adjustable |
| Matériau de la hotte | Painted Steel |
| Finition externe | Painted Steel |
| Certifications/Conformité | CE, REACH, RoHS |
| Modèle | Undercounter |
| Afficher plus |