Learn More
Thermo Scientific™ 384-Well Full-Skirted Plate, Robotic
Tous les produits Thermo Scientific ProduitsDescription
Diamond Ultra PCR plate offers rigidity for reliable performance in high-throughput robotic handling systems. By minimizing warping during thermal cycling and heat sealing, plate dimensions remain consistent and seal integrity remains strong, delivering improved performance in automated PCR and sequencing applications.
Our plastics manufacturing facilities are focused solely on the production of high quality molecular grade plastics. The industry leading manufacturing process does not include any short-cuts and is carried out in world-class facilities run by qualified experts. Our teams of engineers, molecular biologists, and QC/QA managers have the years of experience needed to deliver reliable products that generate accurate and reproducible PCR data, including the Thermo Scientific 384-Well Full-Skirted Plate, Robotic, Barcoded.
Highlights
• Rigid plate for high throughput robotic applications
• Proprietary inert polymer will not bind reaction components
• Working well volume: 25 μL
• Cut Corner: A24
Applications
• PCR
• qPCR/Real-Time PCR
Certifications
Certifié exempt d’ADN humain, de RNase et de DNase.
Spécification
Spécification
| Format | Étalon |
| Array | 16 x 24 |
| Couleur | Transparent |
| Matériau | Virgin Polypropylene |
| Nbre de puits | 384 |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.