missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenascin-R Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 19730-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody is specific to TNR.
Tenascin-R (TNR) is an extracellular matix protein expressed primarily in the central nervous system. It is a member of the tenascin (TN) gene family, which includes at least 3 genes in mammals: TNC (or hexabrachion; MIM 187380), TNX (TNXB; MIM 600985), and TNR (Erickson, 1993). The genes are expressed in distinct tissues at different times during embryonic development and are present in adult tissues.Specifications
| Tenascin-R | |
| Polyclonal | |
| Unconjugated | |
| TNR | |
| Janusin, Restrictin, Tenascin R, TN R, TNR | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 7143 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q92752 | |
| TNR | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction