missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCTN3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16085-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
TCTN3 is a member of the Tectonic protein family, a group of evolutionarily conserved secreted and transmembrane proteins that regulate the Hedgehog (Hh)-mediated patterning of the neural tube. While the expression pattern and function of TCTN3 is not known, it is 58% similar to TCTN1. TCTN1 is expressed during embryonic development in regions that participate in Hh signaling, beginning in the gastrulation stages in the ventral node. Mice expressing mutant TCTN1 die between E13.5 and E16.5 and display holoprosencephaly, a defect associated with reduced Hh signaling, indicating the role of TCTN1 as an Hh activator. At later stages in development, TCTN1 is thought to also act as a repressor on the Hh pathway in the anterior and posterior neural tube.Spécification
TCTN3 | |
Polyclonal | |
Unconjugated | |
TCTN3 | |
C10orf61, TCTN3, TECT3, Tectonic 3, tectonic family member 3, UNQ1881/PRO4324 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26123, 67590 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6NUS6, Q8R2Q6 | |
Tctn3 | |
TCTN3 Fusion Protein Ag9084 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |