missing translation for 'onlineSavingsMsg'
Learn More

TCR beta Armenian Hamster, PE-Cy7, Clone: H57-597, enQuireBio™

Armenian Hamster Monoclonal Antibody

Brand:  EnquireBio QAB79-PE7-100ug

 View more versions of this product

Product Code. 15972498

  • 413.00€ / 100µg

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

TCR beta
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
Armenian Hamster
100 μg
Primary
Store at 2-4°C.
Flow Cytometry
H57-597
PE-Cyanine7
Tcrb
Protein A/G Purified
RUO
21577
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.