Learn More
Huber™ TC100E-F Immersion Cooler
Immersion coolers are a flexible solution for the rapid cooling of liquids and for counter-cooling in heat thermostats or as a substitute for dry ice. With cold traps, evaporation tasks can be realised easily and inexpensively, for example for highly efficient solvent recovery in the laboratory.
Marque:  Huber™ 3005.0107.99
Code nomenclature Nacres: NB.77
Informations supplémentaires : Poids : 59.00000kg
Code produit. 16449705
  • 9497.39€ / 1 pièce
Expédition estimée: 11-10-2024
Description
The TC range of immersion coolers are a flexible solution for a variety of cooling tasks. The devices are simple and universally used for rapid cooling of fluids. Typical applications include counter-cooling for heat circulation thermostats or replacement of dry ice. For many applications immersion coolers are also an environmentally friendly alternative to mains water cooling. The TC immersion coolers are suitable for unsupervised use, for cooling tasks which require continuous cooling or as a variant with temperature control and a Pt100 probe. The models with the type affix -E-have an LED display with entry of nominal values and regulate with a precision of ± 0.5 K. A spiral-shaped or flexible immersion cooling probe made out of stainless steel is available for all models so that fluids can be cooled in virtually all vessel types. The compact housing is made from high quality stainless steel, and is fitted with carrying grips.
Spécification
Spécification
0.16 kW at 0°C, 0.15 kW at -20°C, 0.14 kW at -30°C, 0.13 kW at -50°C, 0.12 kW at -60°C, 0.12 kW at -80°C, 0.07 kW at -90°C, 0.01 kW at -100°C
Cooler
220/240 V
-100°C to 40°C
200 to 240 V, 50/60 Hz