Learn More
STXBP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 11459-1-AP-20UL
Code nomenclature Nacres: NA.46
 Afficher plus de versions de ce produit
Code produit. 16848553
  • 145.00€ / 20µL
Description
This gene encodes a syntaxin-binding protein. The encoded protein appears to play a role in release of neurotransmitters via regulation of syntaxin, a transmembrane attachment protein receptor. Mutations in this gene have been associated with infantile epileptic encephalopathy-4. Alternatively spliced transcript variants have been described.
Spécification
Spécification
STXBP1
Polyclonal
Unconjugated
STXBP1
EIEE4, MUNC18 1, N Sec1, p67, Protein unc 18 homolog 1, Protein unc 18 homolog A, RBSEC1, STXBP1, syntaxin binding protein 1, Unc 18A, UNC18, Unc18 1, UNC18A
Rabbit
Antigen Affinity Chromatography
RUO
20910, 25558, 6812
-20°C
Liquid
Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.46 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O08599, P61764, P61765
Stxbp1
STXBP1 Fusion Protein Ag1978
20 μL
Primary
Human, Rat, Mouse