Learn More
KARTELL Sterile Cylindrical Test Tubes
Click to view available options
Capacité (métrique):
13 ml
Conditionnement:
100 pièces
Description
- Test tubes with quick closure screw caps (PE).
- Leak-proof, and suitable for centrifugation.
- The design of the cap avoids any contamination due to drops of liquid on the inner surface.
- Sterile packing supplied with the screw cap fitted on the test tube.
- Available in: blue (04) - red (10) - yellow (06) - white (03).
Spécification
Spécification
| Capacité (métrique) | 13 ml |
| Longueur (métrique) | 100 mm |
| Matériau | Polyethylene |
| Forme | Cylindrical |
| Stérilité | Stérile |
| Autoclavable | Autoclavable |
| Graduated | No |
| Type de produit | Test Tube |