missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRD5A1 Mouse anti-Human, Clone: 1D5D8, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 66329-1-IG-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Steroid 5alpha-Reductase is an important enzyme in androgen physiology because it catalyzes the conversion of testosterone into the more potent 5alpha-dihydro-testosterone, which mediates androgen effects on target tissues. The enzyme exists as two isoforms: type 1, which is expressed mainly in the skin; and type 2, which is expressed mainly in the prostate. In cultured human skin cells, 5alpha-Reductase 1 shows heterogeneity of protein, and has different levels of transcriptional and translational expression. 5alpha-Reductase 1 is expressed in all portions of the hair follicle, whereas 5alpha-Reductase 2 is expressed only in mesenchymal portions. In addition, 5alpha-Reductase 1 is mainly expressed in human breast carcinoma and may play a role in the in situ production and actions of the potent androgen 5alpha-dihydrotestosterone, including inhibition of cancer cell proliferation in hormone-dependent human breast carcinoma. The 5alpha-Reductase-3alpha-hydroxysteroid dehydrogenase complex is present in the human brain, suggesting that the complex may be involved in the synthesis of neuroactive steroids or the catabolism of neurotoxic steroids.Spécification
| SRD5A1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P18405 | |
| SRD5A1 | |
| SRD5A1 Fusion Protein Ag23351 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence | |
| 1D5D8 | |
| Unconjugated | |
| SRD5A1 | |
| S5AR 1, SR type 1, SRD5A1, Steroid 5 alpha reductase 1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 6715 | |
| -20°C | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu