Learn More
Labconco™ SpillStopper™ Work Surfaces for 5ft. Protector™Premier™Hoods
Keep your equipment safe from spills with the Labconco SpillStopper Work Surface Protector - specifically dished and contoured to conform to the interior liner of Protector Laboratory Hoods.
2227.86€
Code produit | Marque | Type de produit | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Type de produit | Prix | Quantité et disponibilité | |||||
17925012
|
Labconco™
9500501 |
Right Rear |
2227.86€
1 pièce |
Expédition estimée: 08-11-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
SpillStopper work surfaces are dished to contain spills
- The front edge has large radius to aerodynamically direct airflow in hood
- Some surfaces have cutouts for surface-mounted service fixture and/or 3 x 6 in. cup sink
- Flat work surfaces may also be coupled with adjustable height base stand to provide auxiliary workspace
- Service fixture and cup sink are sold separately
Spécification
Epoxy Resin |