missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sorbitol dehydrogenase Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15881-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Sorbitol dehydrogenase catalyzes the interconversion of polyols and their corresponding ketoses, and together with aldose reductase is the reduction of glucose to sorbitol by ALDR1 with NADPH as the cofactor. SORD then oxidizes the sorbitol to fructose using NADcofactor.Spécification
Sorbitol dehydrogenase | |
Polyclonal | |
Unconjugated | |
SORD | |
L iditol 2 dehydrogenase, SDH, Sorbitol dehydrogenase, SORD, SORD1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20322, 24788, 6652 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P27867, Q00796, Q64442 | |
SORD | |
Sorbitol dehydrogenase Fusion Protein Ag8651 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |