Learn More
Dihydrate de tungstate de sodium, purum, Honeywell Fluka™
Catalog Number 15601420
Click to view available options
Quantité:
100 g
500 g
6 x 500 g
10213-10-2
H4Na2O6W
329.846
MFCD00149190
WPZFLQRLSGVIAA-UHFFFAOYSA-N
sodium tungstate dihydrate, sodium tungstate, dihydrate, unii-x1f4ny6u13, tungstic acid, sodium salt, dihydrate, tungstic acid sodium salt dihydrate, tungstic acid sodium salt, disodium dihydrate tungstate, sodium tungstate vi dihydrate, na2o4w.2h2o, na2wo4.2h2o
150191
CHEBI:63939
disodium;dioxido(dioxo)tungsten;dihydrate
O.O.[O-][W](=O)(=O)[O-].[Na+].[Na+]
10213-10-2
H4Na2O6W
329.846
MFCD00149190
WPZFLQRLSGVIAA-UHFFFAOYSA-N
sodium tungstate dihydrate, sodium tungstate, dihydrate, unii-x1f4ny6u13, tungstic acid, sodium salt, dihydrate, tungstic acid sodium salt dihydrate, tungstic acid sodium salt, disodium dihydrate tungstate, sodium tungstate vi dihydrate, na2o4w.2h2o, na2wo4.2h2o
150191
CHEBI:63939
disodium;dioxido(dioxo)tungsten;dihydrate
O.O.[O-][W](=O)(=O)[O-].[Na+].[Na+]
Purum, ≥99 %
  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Nom chimique ou matériau Sodium Tungstate Dihydrate
Quantité 100 g
Formule linéaire Na2WO4 · 2H2O
Numéro UN NONH for all modes of transport
Poids de la formule 329.85g/mol
Pourcentage de pureté ≥99%
Qualité purum