Learn More
Brady™ Small Core Pre-printed GHS CLP Labels
Supplied with pre-printed red diamonds, ready for you to print the required warning, compliant to the latest regulations.
Marque: Brady™ 622339
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
For you to print the required warning, compliant to the latest regulations.
Spécification
Pre-printed Desktop Printer Labels for CLP-GHS Hazardous Substances | |
None | |
Matte | |
Front | |
Labels | |
3 Stacked Red Diamonds in Pyramid Formation per Sticker | |
BBP11, BBP12, PR Plus, THT 25 mm Core Printers | |
-40°C | |
200 Labels/Roll | |
Rectangle | |
-40°F to +122°F | |
0.0032 in. | |
105.00 mm (W) x 155.00 mm (H) | |
155.00 mm | |
105.00 mm | |
105.00 mm |
Permanent Acrylic | |
Red on White | |
Paper | |
Thermal Transfer | |
Non-sterile | |
Hazard communication identification | |
1 | |
R7950 | |
200 Stickers | |
105.00 mm (W) x 155.00 mm (H) | |
-40°C to +50°C | |
0.081 mm | |
6.1 in. | |
4.13 in. | |
105.00 mm |