Learn More
Brady™ Small Core Pre-printed GHS CLP Labels
Supplied with pre-printed red diamonds, ready for you to print the required warning, compliant to the latest regulations.
Marque: Brady™ 622322
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
For you to print the required warning, compliant to the latest regulations.
Spécification
UL Recognized | |
Permanent Acrylic | |
Red on White | |
Polypropylene | |
Thermal Transfer | |
Non-sterile | |
Hazard communication identification | |
1 | |
R6000HF | |
300 Stickers | |
75.00 mm (W) x 110.00 mm (H) | |
-20°C to +100°C | |
0.079 mm | |
4.33 in. | |
2.95 in. | |
75.00 mm |
Pre-printed Desktop Printer Labels for CLP-GHS Hazardous Substances | |
None | |
Gloss | |
Front | |
Labels | |
3 Red Diamonds per Sticker | |
BBP11, BBP12, PR Plus, THT 25 mm Core Printers | |
-20°C | |
300 Labels/Roll | |
Rectangle | |
-4°F to 212°F | |
0.0031 in. | |
75.00 mm (W) x 110.00 mm (H) | |
110.00 mm | |
75.00 mm | |
75.00 mm |