missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SFTPC Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA542014
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Peptide sequence: MDVGSKEVLM ESPPDYSAAP RGRFGIPCCP VHLKRLLIVV VVVVLIVVVI Sequence homology: Dog: 100%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 79%; Rabbit: 100%; Rat: 100%; Sheep: 85%.
This gene encodes the pulmonary-associated surfactant protein C (SPC), an extremely hydrophobic surfactant protein essential for lung function and homeostasis after birth. Pulmonary surfactant is a surface-active lipoprotein complex composed of 90% lipids and 10% proteins which include plasma proteins and apolipoproteins SPA, SPB, SPC and SPD. The surfactant is secreted by the alveolar cells of the lung and maintains the stability of pulmonary tissue by reducing the surface tension of fluids that coat the lung. Multiple mutations in this gene have been identified, which cause pulmonary surfactant metabolism dysfunction type 2, also called pulmonary alveolar proteinosis due to surfactant protein C deficiency, and are associated with interstitial lung disease in older infants, children, and adults. Alternatively spliced transcript variants encoding different protein isoforms have been identified.
Spécification
| SFTPC | |
| Polyclonal | |
| Unconjugated | |
| Sftpc | |
| BRICD6; BRICHOS domain containing 6; LOW QUALITY PROTEIN: pulmonary surfactant-associated protein C; pro-SpC; PSP C; PSP-C; pulmonary surfactant apoprotein-2 SP-C; pulmonary surfactant associated protein C; pulmonary surfactant protein C; pulmonary surfactant-associated protein C; Pulmonary surfactant-associated proteolipid SPL(Val); SFTP 2; SFTP2; Sftp-2; Sftpc; SMDP2; SP C; SP5; SPC; SP-C; surfactant associated protein C; surfactant protein C; Surfactant pulmonary-associated protein C; Surfactant, pulmonary-associated protein C; surfactant-associated protein; type I SP-C; type I surfactant protein C; type II SP-C; type II surfactant protein C | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 6440 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot, Surface Plasmon Resonance | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P11686 | |
| Sftpc | |
| Synthetic peptide directed towards the N-terminal of human SFTPC (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu