missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One Tubes de sérum : Activateur de coagulation avec séparateur de gel
For routine clinical chemistry, immunohematology, serology or immunology testing. Greiner Bio-One VACUETTE™ Z Serum Sep Clot Activator Tubes - Non-ridged (pull cap) are clear as glass with equal or better performance.
Marque: Greiner Bio-One 454228
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
- Coated with micronized silica particles which activate clotting when tubes are gently inverted
- Tubes with gel contain a barrier gel on the tube bottom that during centrifugation moves upward to the serum-clot interface to form a stable barrier, separating serum from cells
- Molded to be virtually unbreakable, providing a better level of safety than glass
- Tubes have an 18-month shelf life from the date of manufacture
- Pediatric tubes come with white rings for quick identification
- Safety Twist Caps can be removed and replaced several times
- Nonscrew caps can be removed and replaced as often as required
Spécification
Microscopic Silica Particles with Separation Gel | |
Non-ridged Pull Cap | |
Gold with Gold Ring |
Plastic | |
3.5 mL | |
13 x 75 mm |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu